Assay Details
| Gene Symbol: Prpf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177806.3 | NP_808474.2 | 5 - 6 | 890 | 147 | |
| GenBank mRNA | AK154170.1 | - | 5 - 6 | 1414 | 147 | |
| AK168462.1 | - | 5 - 6 | 884 | 147 | ||
| BC014689.1 | - | 5 - 6 | 869 | 147 | ||
| BC030904.1 | - | 4 - 5 | 709 | 147 | ||
| BC034078.1 | - | 7 - 8 | 1101 | 147 | ||
| BC076622.1 | - | 4 - 5 | 476 | 147 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154170.1 transcripts.
Set Membership: |
|

Prpf39
Genomic Map