Assay Details
| Gene Symbol: Prpf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177806.3 | NP_808474.2 | 7 - 8 | 1166 | 172 | |
| GenBank mRNA | AK154170.1 | - | 7 - 8 | 1690 | 172 | |
| BC014689.1 | - | 7 - 8 | 1145 | 172 | ||
| BC030904.1 | - | 6 - 7 | 985 | 172 | ||
| BC033575.1 | - | 3 - 4 | 2916 | 172 | ||
| BC034078.1 | - | 9 - 10 | 1377 | 172 | ||
| BC076622.1 | - | 6 - 7 | 752 | 172 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154170.1 transcripts.
Set Membership: |
|

Prpf39
Genomic Map