Assay Details
| Gene Symbol: Pla2g4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144750.1 | - | 2 - 3 | 151 | 94 | |
| AK172121.1 | - | 2 - 3 | 191 | 94 | ||
| BC003816.1 | - | 2 - 3 | 191 | 3485483 | 94 | |
| M72394.1 | - | 2 - 3 | 139 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144750.1, BC003816.1, M72394.1 transcripts.
Set Membership: |
|

Pla2g4a
Genomic Map