Assay Details
| Gene Symbol: Fmo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144878.1 | NP_659127.1 | 6 - 7 | 845 | 141 | |
| GenBank mRNA | AF461145.1 | - | 6 - 7 | 845 | 141 | |
| BC106858.1 | - | 5 - 6 | 836 | 40047236 | 141 | |
| BC106859.1 | - | 5 - 6 | 836 | 40047237 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fmo4
Genomic Map