Assay Details
| Gene Symbol: Trmt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026876.3 | NP_081152.2 | 12 - 13 | 1972 | 105 | |
| GenBank mRNA | AK004493.1 | - | 4 - 5 | 465 | 105 | |
| AK146088.1 | - | 12 - 13 | 1973 | 105 | ||
| BC022682.1 | - | 7 - 8 | 1008 | 105 | ||
| BC037478.1 | - | 8 - 9 | 1152 | 105 | ||
| BC064024.1 | - | 12 - 13 | 1770 | 105 | ||
| BC110425.1 | - | 12 - 13 | 1955 | 5369595 | 105 | |
| BC132299.1 | - | 12 - 13 | 1830 | 40130576 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt1l
Genomic Map