Assay Details
| Gene Symbol: Trmt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026876.3 | NP_081152.2 | 2 - 3 | 568 | 70 | |
| GenBank mRNA | AK146088.1 | - | 2 - 3 | 569 | 70 | |
| AK148524.1 | - | 2 - 3 | 675 | 70 | ||
| BC064024.1 | - | 2 - 3 | 338 | 70 | ||
| BC110425.1 | - | 2 - 3 | 551 | 5369595 | 70 | |
More Information
Set Membership: |
|

Trmt1l
Genomic Map