Assay Details
| Gene Symbol: Trmt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026876.3 | NP_081152.2 | 6 - 7 | 999 | 123 | |
| GenBank mRNA | AK146088.1 | - | 6 - 7 | 1000 | 123 | |
| AK148524.1 | - | 6 - 7 | 1106 | 123 | ||
| BC037478.1 | - | 2 - 3 | 179 | 123 | ||
| BC061491.1 | - | 6 - 7 | 1005 | 123 | ||
| BC064024.1 | - | 6 - 7 | 769 | 123 | ||
| BC110425.1 | - | 6 - 7 | 982 | 5369595 | 123 | |
| BC132299.1 | - | 6 - 7 | 857 | 40130576 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt1l
Genomic Map