Assay Details
| Gene Symbol: Zcchc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122675.1 | NP_001116147.1 | 10 - 11 | 2099 | 111 | |
| NM_001122676.1 | NP_001116148.1 | 10 - 11 | 2099 | 111 | ||
| GenBank mRNA | AK037100.1 | - | 4 - 5 | 272 | 111 | |
| AK173248.1 | - | 10 - 11 | 1816 | 111 | ||
| BC035528.1 | - | 7 - 8 | 589 | 111 | ||
| BC082306.1 | - | 6 - 7 | 507 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zcchc2
Genomic Map