Assay Details
| Gene Symbol: Lemd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033250.4 | NP_001028422.2 | 2 - 3 | 175 | 63 | |
| GenBank mRNA | AK016010.1 | - | 2 - 3 | 180 | 63 | |
| AK144269.1 | - | 182 | 63 | |||
| BC048759.1 | - | 2 - 3 | 139 | 63 | ||
| BC115462.1 | - | 2 - 3 | 156 | 40100287 | 63 | |
| BC115463.1 | - | 2 - 3 | 156 | 40100292 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lemd1
Genomic Map