Assay Details
| Gene Symbol: Srgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081011.2 | NP_001074480.2 | 14 - 15 | 2273 | 73 | |
| GenBank mRNA | AK161932.1 | - | 3 - 4 | 177 | 73 | |
| AY057900.1 | - | 5 - 6 | 262 | 73 | ||
| BC030457.1 | - | 13 - 14 | 1580 | 73 | ||
| BC151081.1 | - | 13 - 14 | 1891 | 9087994 | 73 | |
| BC151082.1 | - | 13 - 14 | 1891 | 9087995 | 73 | |
| BC158055.1 | - | 13 - 14 | 1889 | 9088163 | 73 | |
| BC172152.1 | - | 13 - 14 | 1891 | 9054822 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK161932.1 transcripts.
Set Membership: |
|

Srgap2
Genomic Map