Assay Details
| Gene Symbol: Srgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081011.2 | NP_001074480.2 | 9 - 10 | 1876 | 66 | |
| GenBank mRNA | BC030457.1 | - | 8 - 9 | 1183 | 66 | |
| BC151081.1 | - | 8 - 9 | 1494 | 9087994 | 66 | |
| BC151082.1 | - | 8 - 9 | 1494 | 9087995 | 66 | |
| BC158055.1 | - | 8 - 9 | 1492 | 9088163 | 66 | |
| BC172152.1 | - | 8 - 9 | 1494 | 9054822 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srgap2
Genomic Map