Assay Details
| Gene Symbol: Srgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081011.2 | NP_001074480.2 | 10 - 11 | 2074 | 92 | |
| GenBank mRNA | AK186803.1 | - | 110 | 92 | ||
| AY057900.1 | - | 1 - 2 | 63 | 92 | ||
| BC030457.1 | - | 9 - 10 | 1381 | 92 | ||
| BC151081.1 | - | 9 - 10 | 1692 | 9087994 | 92 | |
| BC151082.1 | - | 9 - 10 | 1692 | 9087995 | 92 | |
| BC158055.1 | - | 9 - 10 | 1690 | 9088163 | 92 | |
| BC172152.1 | - | 9 - 10 | 1692 | 9054822 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srgap2
Genomic Map