Assay Details
| Gene Symbol: Dtymk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105667.1 | NP_001099137.1 | 2 - 3 | 341 | 66 | |
| GenBank mRNA | AK088416.1 | - | 2 - 3 | 301 | 66 | |
| BC030178.1 | - | 2 - 3 | 303 | 5039765 | 66 | |
| CB182101.1 | - | 2 - 3 | 297 | 66 | ||
| S82853.1 | - | 2 - 3 | 282 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dtymk
Genomic Map