Assay Details
| Gene Symbol: Dtymk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105667.1 | NP_001099137.1 | 3 - 4 | 434 | 84 | |
| GenBank mRNA | AK009220.1 | - | 3 - 4 | 407 | 84 | |
| AK088416.1 | - | 3 - 4 | 394 | 84 | ||
| BC030178.1 | - | 3 - 4 | 396 | 5039765 | 84 | |
| CB182101.1 | - | 3 - 4 | 390 | 84 | ||
| S82853.1 | - | 3 - 4 | 375 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dtymk
Genomic Map