Assay Details
| Gene Symbol: Dtymk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105667.1 | NP_001099137.1 | 4 - 5 | 637 | 87 | |
| GenBank mRNA | AK009220.1 | - | 4 - 5 | 610 | 87 | |
| AK088416.1 | - | 4 - 5 | 597 | 87 | ||
| AK205785.1 | - | 1 - 2 | 161 | 87 | ||
| AK207877.1 | - | 1 - 2 | 68 | 87 | ||
| BC030178.1 | - | 4 - 5 | 599 | 5039765 | 87 | |
| CB182101.1 | - | 4 - 5 | 593 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dtymk
Genomic Map