Assay Details
| Gene Symbol: Nudt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153561.3 | NP_705789.1 | 3 - 4 | 520 | 114 | |
| GenBank mRNA | BC027267.1 | - | 3 - 4 | 514 | 3500531 | 114 |
More Information
Set Membership: |
|

Nudt6
Genomic Map