Assay Details
| Gene Symbol: Adad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009350.3 | NP_033376.2 | 1 - 2 | 180 | 109 | |
| GenBank mRNA | BC107346.2 | - | 1 - 2 | 158 | 109 | |
| X84693.1 | - | 1 - 2 | 109 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Adad1
Genomic Map