Assay Details
| Gene Symbol: Adad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009350.3 | NP_033376.2 | 3 - 4 | 549 | 84 | |
| GenBank mRNA | BC107346.2 | - | 3 - 4 | 527 | 84 | |
| X84693.1 | - | 3 - 4 | 537 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adad1
Genomic Map