Assay Details
| Gene Symbol: Adad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009350.3 | NP_033376.2 | 6 - 7 | 916 | 154 | |
| GenBank mRNA | AK133008.1 | - | 3 - 4 | 332 | 154 | |
| X84693.1 | - | 6 - 7 | 904 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adad1
Genomic Map