Assay Details
| Gene Symbol: Adad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009350.3 | NP_033376.2 | 7 - 8 | 1033 | 126 | |
| GenBank mRNA | X84693.1 | - | 7 - 8 | 1021 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adad1
Genomic Map