Assay Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172516.4 | NP_766104.2 | 10 - 11 | 2495 | 65 | |
| GenBank mRNA | AK050542.1 | - | 6 - 7 | 741 | 65 | |
| AK083696.1 | - | 10 - 11 | 2393 | 65 | ||
| AK132119.1 | - | 10 - 11 | 2370 | 65 | ||
| AK145449.1 | - | 10 - 11 | 2340 | 65 | ||
| AK172954.1 | - | 10 - 11 | 2341 | 65 | ||
| AY208852.1 | - | 10 - 11 | 2342 | 65 | ||
| AY429676.1 | - | 10 - 11 | 2381 | 65 | ||
| BC019133.1 | - | 8 - 9 | 1130 | 65 | ||
| BC089380.1 | - | 7 - 8 | 912 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dstyk
Genomic Map