Assay Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172516.4 | NP_766104.2 | 1 - 2 | 414 | 86 | |
| GenBank mRNA | AK020880.1 | - | 1 - 2 | 300 | 86 | |
| AK083696.1 | - | 1 - 2 | 312 | 86 | ||
| AK132119.1 | - | 1 - 2 | 289 | 86 | ||
| AK145449.1 | - | 1 - 2 | 259 | 86 | ||
| AK172954.1 | - | 1 - 2 | 261 | 86 | ||
| AY208852.1 | - | 1 - 2 | 261 | 86 | ||
| AY429676.1 | - | 1 - 2 | 300 | 86 | ||
| BC130226.1 | - | 1 - 2 | 290 | 40141770 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dstyk
Genomic Map