Assay Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172516.4 | NP_766104.2 | 8 - 9 | 2258 | 61 | |
| GenBank mRNA | AK050542.1 | - | 4 - 5 | 504 | 61 | |
| AK083696.1 | - | 8 - 9 | 2156 | 61 | ||
| AK132119.1 | - | 8 - 9 | 2133 | 61 | ||
| AK145449.1 | - | 8 - 9 | 2103 | 61 | ||
| AK172954.1 | - | 8 - 9 | 2104 | 61 | ||
| AY208852.1 | - | 8 - 9 | 2105 | 61 | ||
| AY429676.1 | - | 8 - 9 | 2144 | 61 | ||
| BC019133.1 | - | 6 - 7 | 893 | 61 | ||
| BC089380.1 | - | 5 - 6 | 675 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dstyk
Genomic Map