Assay Details
| Gene Symbol: Adam19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291890.1 | NP_001278819.1 | 12 - 13 | 1256 | 69 | |
| NM_001291891.1 | NP_001278820.1 | 12 - 13 | 1451 | 69 | ||
| NM_009616.4 | NP_033746.1 | 12 - 13 | 1441 | 69 | ||
| GenBank mRNA | AF019887.2 | - | 12 - 13 | 1346 | 69 | |
| AK147217.1 | - | 12 - 13 | 1364 | 69 | ||
| AK147549.1 | - | 12 - 13 | 1257 | 69 | ||
| BC093480.1 | - | 12 - 13 | 1442 | 69 | ||
| D50410.2 | - | 12 - 13 | 1373 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam19
Genomic Map