Assay Details
| Gene Symbol: Lrrfip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111311.1 | NP_001104781.1 | 5 - 6 | 705 | 73 | |
| NM_001111312.1 | NP_001104782.1 | 6 - 7 | 664 | 73 | ||
| NM_008515.4 | NP_032541.1 | 14 - 15 | 1221 | 73 | ||
| GenBank mRNA | AF045573.1 | - | 14 - 15 | 1158 | 73 | |
| AK041162.1 | - | 6 - 7 | 615 | 73 | ||
| AK044174.1 | - | 11 - 12 | 884 | 73 | ||
| AK077637.1 | - | 5 - 6 | 710 | 73 | ||
| AK133661.1 | - | 5 - 6 | 686 | 73 | ||
| AK134120.1 | - | 5 - 6 | 697 | 73 | ||
| BC144955.1 | - | 5 - 6 | 558 | 73 | ||
| BC144956.1 | - | 16 - 17 | 1179 | 9053504 | 73 | |
| BC145642.1 | - | 16 - 17 | 1179 | 40130945 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrfip1
Genomic Map