Assay Details
| Gene Symbol: Ralgps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159965.1 | NP_001153437.1 | 14 - 15 | 1485 | 68 | |
| NM_001159966.1 | NP_001153438.1 | 14 - 15 | 1383 | 68 | ||
| NM_001159968.1 | NP_001153440.1 | 13 - 14 | 1380 | 68 | ||
| NM_023884.4 | NP_076373.3 | 14 - 15 | 1425 | 68 | ||
| GenBank mRNA | AF312924.1 | - | 13 - 14 | 1333 | 68 | |
| AK018622.1 | - | 14 - 15 | 1427 | 68 | ||
| AK019543.1 | - | 13 - 14 | 1383 | 68 | ||
| AK029049.1 | - | 14 - 15 | 1426 | 68 | ||
| AK036803.1 | - | 13 - 14 | 1357 | 68 | ||
| AK165002.1 | - | 14 - 15 | 1377 | 68 | ||
| BC052663.1 | - | 14 - 15 | 1402 | 30051209 | 68 | |
More Information
Set Membership: |
|

Ralgps2
Genomic Map