Assay Details
| Gene Symbol: Gm4926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028066.1 | 2 - 3 | 388 | 85 | ||
| GenBank mRNA | AK079066.1 | - | 2 - 3 | 389 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | developmental pluripotency associated 1 | Dppa1 | C77727 | -7.8kb | Sense | Chr. 11 - 46607010 - 46632233 on NCBI Build GRCm38 |
| Downstream | T cell immunoglobulin and mucin domain containing 2 | Timd2 |
AA816106, AU019358, AU019662, C78111, G9D3, TIM-2, Tim2 |
27.7kb | Sense | Chr. 11 - 46668960 - 46707104 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm4926
Genomic Map