Assay Details
| Gene Symbol: 4932429P19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016536.1 | - | 1 | 3254 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 71 like | Ccdc71l |
2010109K11Rik, AV375874 |
-45.7kb | Anti-sense | Chr. 12 - 32378789 - 32382943 on NCBI Build GRCm38 |
| Downstream | nicotinamide phosphoribosyltransferase | Nampt |
1110035O14Rik, AI314458, AI480535, NAmPRTase, Pbef, Pbef1, Visfatin |
391.3kb | Anti-sense | Chr. 12 - 32820335 - 32853369 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4932429P19Rik
Genomic Map