Assay Details
| Gene Symbol: Grhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161406.1 | NP_001154878.1 | 2 - 3 | 361 | 63 | |
| GenBank mRNA | AF411212.1 | - | 2 - 3 | 332 | 63 | |
| AK076307.1 | - | 2 - 3 | 346 | 63 | ||
| AK161048.1 | - | 2 - 3 | 243 | 63 | ||
| AK167035.1 | - | 2 - 3 | 361 | 63 | ||
| BC013080.1 | - | 2 - 3 | 301 | 3499860 | 63 | |
| BC037233.1 | - | 2 - 3 | 353 | 4951512 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grhl1
Genomic Map