Assay Details
| Gene Symbol: Neu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015750.3 | NP_056565.1 | 1 - 2 | 69 | 81 | |
| GenBank mRNA | AB028023.1 | - | 1 - 2 | 71 | 81 | |
| BC105657.1 | - | 1 - 2 | 71 | 40046586 | 81 | |
| BC105658.1 | - | 1 - 2 | 71 | 40046587 | 81 | |
| BC105659.1 | - | 1 - 2 | 71 | 40046588 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB028023.1, BC105657.1, BC105658.1, BC105659.1, NM_015750.3 transcripts.
Set Membership: |
|

Neu2
Genomic Map