Assay Details
| Gene Symbol: Mgst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025569.1 | NP_079845.1 | 1 - 2 | 47 | 78 | |
| GenBank mRNA | AK003246.1 | - | 1 - 2 | 47 | 78 | |
| AK003309.1 | - | 1 - 2 | 33 | 78 | ||
| AK003476.1 | - | 1 - 2 | 40 | 78 | ||
| AK003492.1 | - | 1 - 2 | 40 | 78 | ||
| AK008533.1 | - | 1 - 2 | 36 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003246.1,AK003309.1,AK003476.1,AK003492.1,AK008533.1,NM_025569.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mgst3
Genomic Map