Assay Details
| Gene Symbol: Mgst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025569.1 | NP_079845.1 | 4 - 5 | 304 | 53 | |
| GenBank mRNA | AK003246.1 | - | 4 - 5 | 304 | 53 | |
| AK003309.1 | - | 4 - 5 | 290 | 53 | ||
| AK003476.1 | - | 4 - 5 | 297 | 53 | ||
| AK003492.1 | - | 4 - 5 | 297 | 53 | ||
| AK008533.1 | - | 4 - 5 | 293 | 53 | ||
| AK135742.1 | - | 4 - 5 | 279 | 53 | ||
| BC029669.1 | - | 3 - 4 | 283 | 5372427 | 53 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mgst3
Genomic Map