Assay Details
| Gene Symbol: 4930412E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015118.1 | - | 1 - 2 | 374 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribonuclease P RNA-like 2 | Rprl2 |
RPRH3, Rmrp3 |
-1.2mb | Sense | Chr. 3 - 22251370 - 22251607 on NCBI Build GRCm38 |
| Downstream | N-acetylated alpha-linked acidic dipeptidase-like 2 | Naaladl2 |
2810043G22Rik, EG635702, Gm1021 |
336.5kb | Anti-sense | Chr. 3 - 23798103 - 24784006 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930412E21Rik
Genomic Map