Assay Details
| Gene Symbol: 4930412E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015118.1 | - | 2 - 3 | 441 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribonuclease P RNA-like 2 | Rprl2 |
RPRH3, Rmrp3 |
-1.2mb | Sense | Chr. 3 - 22251370 - 22251607 on NCBI Build GRCm38 |
| Downstream | N-acetylated alpha-linked acidic dipeptidase-like 2 | Naaladl2 |
2810043G22Rik, EG635702, Gm1021 |
276.7kb | Anti-sense | Chr. 3 - 23798103 - 24784006 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058245 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672125 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760442 | AK053602.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760442 | XM_893895.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760442 | XM_912438.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787002 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537932 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545320 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 661882 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714042 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233563 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635199 | AK017438.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635199 | XM_896507.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635199 | XM_919110.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441954 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885870 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 194493 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121508 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8344 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17338 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058245 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04846252 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04846209 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04846195 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04846232 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04846224 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112822 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04846209 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04846232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02746815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02505919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03108669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02789397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02991503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03182815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02642777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03182807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03083958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03182812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02789419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02991513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02689329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21991663 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103870050 | - | - |

4930412E21Rik
Genomic Map