Assay Details
| Gene Symbol: Lims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193303.1 | NP_001180232.1 | 1 - 2 | 232 | 75 | |
| GenBank mRNA | AK009390.1 | - | 1 - 2 | 233 | 75 | |
| DQ003608.1 | - | 1 - 2 | 217 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lims1
Genomic Map