Assay Details
| Gene Symbol: Lims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193303.1 | NP_001180232.1 | 4 - 5 | 571 | 64 | |
| NM_026148.3 | NP_080424.2 | 4 - 5 | 565 | 64 | ||
| NM_201242.1 | NP_957694.1 | 4 - 5 | 589 | 64 | ||
| GenBank mRNA | AK009390.1 | - | 4 - 5 | 572 | 64 | |
| AK082932.1 | - | 4 - 5 | 434 | 64 | ||
| AK088939.1 | - | 4 - 5 | 440 | 64 | ||
| AK151235.1 | - | 4 - 5 | 440 | 64 | ||
| AK151938.1 | - | 4 - 5 | 432 | 64 | ||
| AK152679.1 | - | 4 - 5 | 432 | 64 | ||
| AK153346.1 | - | 4 - 5 | 440 | 64 | ||
| BC005621.1 | - | 4 - 5 | 420 | 3711611 | 64 | |
| BC067393.1 | - | 4 - 5 | 363 | 64 | ||
| DQ003608.1 | - | 4 - 5 | 556 | 64 | ||
| EU122953.1 | - | 4 - 5 | 555 | 64 | ||
| EU122954.1 | - | 4 - 5 | 423 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lims1
Genomic Map