Assay Details
| Gene Symbol: Cd55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010016.2 | NP_034146.2 | 1 - 2 | 171 | 135 | |
| GenBank mRNA | AK030285.1 | - | 1 - 2 | 163 | 135 | |
| AK160994.1 | - | 1 - 2 | 149 | 135 | ||
| BC011314.1 | - | 1 - 2 | 126 | 3663561 | 135 | |
| BU515742.1 | - | 1 - 2 | 175 | 135 | ||
| CT010328.1 | - | 1 - 2 | 100 | 135 | ||
| L41366.1 | - | 1 - 2 | 115 | 135 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cd55
Genomic Map