Assay Details
| Gene Symbol: Cd55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010016.2 | NP_034146.2 | 7 - 8 | 1077 | 77 | |
| GenBank mRNA | AK160994.1 | - | 7 - 8 | 1055 | 77 | |
| BC011314.1 | - | 7 - 8 | 1032 | 3663561 | 77 | |
| CT010328.1 | - | 7 - 8 | 1006 | 77 | ||
| D63679.1 | - | 7 - 8 | 988 | 77 | ||
| L41366.1 | - | 7 - 8 | 1021 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd55
Genomic Map