Assay Details
| Gene Symbol: Mfsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114662.1 | NP_001108134.1 | 7 - 8 | 1459 | 71 | |
| NM_172510.4 | NP_766098.2 | 7 - 8 | 1459 | 71 | ||
| GenBank mRNA | AK038882.1 | - | 7 - 8 | 1352 | 71 | |
| AK044667.1 | - | 7 - 8 | 1333 | 71 | ||
| AK048692.1 | - | 6 - 7 | 1261 | 71 | ||
| AK085484.1 | - | 4 - 5 | 875 | 71 | ||
| AK162495.1 | - | 7 - 8 | 1363 | 71 | ||
| BC058790.1 | - | 7 - 8 | 1308 | 6395070 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd4
Genomic Map