Assay Details
| Gene Symbol: Mfsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114662.1 | NP_001108134.1 | 8 - 9 | 1522 | 76 | |
| NM_172510.4 | NP_766098.2 | 8 - 9 | 1522 | 76 | ||
| GenBank mRNA | AK038882.1 | - | 8 - 9 | 1415 | 76 | |
| AK044667.1 | - | 8 - 9 | 1396 | 76 | ||
| AK048692.1 | - | 7 - 8 | 1324 | 76 | ||
| AK085484.1 | - | 5 - 6 | 938 | 76 | ||
| AK162495.1 | - | 8 - 9 | 1426 | 76 | ||
| BC058790.1 | - | 8 - 9 | 1371 | 6395070 | 76 | |
| FN434126.1 | - | 1 - 2 | 55 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd4
Genomic Map