Assay Details
| Gene Symbol: Sgcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011891.4 | NP_036021.1 | 5 - 6 | 543 | 87 | |
| GenBank mRNA | AB024923.1 | - | 5 - 6 | 764 | 87 | |
| AK040974.1 | - | 5 - 6 | 545 | 87 | ||
| AK047384.1 | - | 1285 | 87 | |||
| BC130019.1 | - | 5 - 6 | 533 | 40140624 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgcd
Genomic Map