Assay Details
| Gene Symbol: Rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028862.3 | NP_083138.2 | 1 - 2 | 317 | 141 | |
| GenBank mRNA | AK014408.1 | - | 1 - 2 | 318 | 141 | |
| AK043002.1 | - | 1 - 2 | 304 | 141 | ||
| BB618481.1 | - | 1 - 2 | 184 | 141 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014408.1, AK043002.1, NM_028862.3 transcripts.
Set Membership: |
|

Rnf145
Genomic Map