Assay Details
| Gene Symbol: Rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166553.1 | NP_001160025.1 | 2 - 3 | 501 | 115 | |
| NM_028862.3 | NP_083138.2 | 2 - 3 | 520 | 115 | ||
| GenBank mRNA | AK014408.1 | - | 2 - 3 | 521 | 115 | |
| AK043002.1 | - | 2 - 3 | 507 | 115 | ||
| AK087359.1 | - | 2 - 3 | 437 | 115 | ||
| AK139310.1 | - | 2 - 3 | 418 | 115 | ||
| BB618481.1 | - | 2 - 3 | 387 | 115 | ||
| BC040799.1 | - | 1 - 2 | 196 | 3988157 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf145
Genomic Map