Assay Details
| Gene Symbol: Rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166553.1 | NP_001160025.1 | 4 - 5 | 713 | 69 | |
| NM_028862.3 | NP_083138.2 | 4 - 5 | 732 | 69 | ||
| GenBank mRNA | AK043002.1 | - | 4 - 5 | 719 | 69 | |
| AK087359.1 | - | 4 - 5 | 649 | 69 | ||
| AK139310.1 | - | 4 - 5 | 630 | 69 | ||
| BC040799.1 | - | 3 - 4 | 408 | 3988157 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf145
Genomic Map