| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100003425.mm.1 |
NM_028862.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003425.mm.1 |
NM_028862.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003425.mm.1 |
TC1100000440.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100003423.mm.1 |
NM_001166553.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003423.mm.1 |
NM_001166553.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003423.mm.1 |
NM_028862.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003423.mm.1 |
NM_028862.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003423.mm.1 |
TC1100000440.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100003424.mm.1 |
NM_001166553.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003424.mm.1 |
NM_001166553.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003424.mm.1 |
TC1100000440.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100003422.mm.1 |
NM_001166553.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003422.mm.1 |
NM_001166553.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003422.mm.1 |
NM_028862.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003422.mm.1 |
NM_028862.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003422.mm.1 |
TC1100000440.mm.1 |
MTA TC |