| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100003431.mm.1 |
NM_028862.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003431.mm.1 |
NM_028862.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003431.mm.1 |
TC1100000440.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100003430.mm.1 |
NM_028862.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003430.mm.1 |
NM_028862.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003430.mm.1 |
TC1100000440.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100003427.mm.1 |
NM_028862.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003427.mm.1 |
NM_028862.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003427.mm.1 |
TC1100000440.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100003428.mm.1 |
NM_028862.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003428.mm.1 |
NM_028862.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003428.mm.1 |
TC1100000440.mm.1 |
MTA TC |