Assay Details
| Gene Symbol: Rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028862.3 | NP_083138.2 | 9 - 10 | 1612 | 92 | |
| GenBank mRNA | AK014408.1 | - | 9 - 10 | 1613 | 92 | |
| AK043002.1 | - | 9 - 10 | 1599 | 92 | ||
| BC040799.1 | - | 8 - 9 | 1288 | 3988157 | 92 | |
| BC051064.1 | - | 2 - 3 | 315 | 92 | ||
| BC055485.1 | - | 3 - 4 | 396 | 92 | ||
More Information
Set Membership: |
|

Rnf145
Genomic Map