Assay Details
| Gene Symbol: Rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166553.1 | NP_001160025.1 | 1 - 2 | 293 | 89 | |
| GenBank mRNA | AK087359.1 | - | 1 - 2 | 229 | 89 | |
| AK139310.1 | - | 1 - 2 | 210 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087359.1, NM_001166553.1 transcripts.
Set Membership: |
|

Rnf145
Genomic Map