Assay Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148673.3 | NP_683514.2 | 12 - 13 | 1313 | 59 | |
| NM_198936.1 | NP_945174.1 | 11 - 12 | 1234 | 59 | ||
| GenBank mRNA | AK029117.1 | - | 11 - 12 | 1204 | 59 | |
| AK046262.1 | - | 11 - 12 | 1205 | 59 | ||
| AK047597.1 | - | 12 - 13 | 1301 | 59 | ||
| AK049178.1 | - | 11 - 12 | 1203 | 59 | ||
| BC013810.1 | - | 9 - 10 | 833 | 59 | ||
| BC023057.1 | - | 1 - 2 | 88 | 59 | ||
| BC060954.1 | - | 11 - 12 | 1202 | 59 | ||
| BC082780.1 | - | 12 - 13 | 1293 | 59 | ||
| BC106099.1 | - | 11 - 12 | 1215 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slu7
Genomic Map