Assay Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148673.3 | NP_683514.2 | 8 - 9 | 870 | 92 | |
| NM_198936.1 | NP_945174.1 | 7 - 8 | 791 | 92 | ||
| GenBank mRNA | AK029117.1 | - | 7 - 8 | 761 | 92 | |
| AK046262.1 | - | 7 - 8 | 762 | 92 | ||
| AK047597.1 | - | 8 - 9 | 858 | 92 | ||
| AK049178.1 | - | 7 - 8 | 760 | 92 | ||
| BC060954.1 | - | 7 - 8 | 759 | 92 | ||
More Information
Set Membership: |
|

Slu7
Genomic Map